3-Amino-5-methoxypicolinic acid ≥98%

Lieferant: Apollo Scientific

OR510182-100MG
APOSOR510182-100MGEA 344 CHF
APOSOR510182-100MG
3-Amino-5-methoxypicolinic acid ≥98%
3-Amino-5-methoxypicolinic acid
Formel: C₇H₈N₂O₃
MW: 168.15 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD22544090
CAS-Nummer: 1393585-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR