6-Aminonicotinsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyms: 6-Aminopyridine-3-carboxylic acid, 6-Amino-3-pyridinecarboxylic acid

OR5070-100G
APOSOR5070-100GEA 69.6 CHF
APOSOR5070-100G
6-Aminonicotinsäure 98%
6-Aminonicotinsäure
Formel: C₆H₆N₂O₂
Melting Pt: 320 °C (dec.)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00006326
CAS-Nummer: 3167-49-5
EINECS: 221-630-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR