(2R)-2-Amino-4-(methylamino)-4-oxo-butanoic acid

Lieferant: Apollo Scientific
Achtung

OR480363-1G
APOSOR480363-1GEA 244 CHF
APOSOR480363-1G
(2R)-2-Amino-4-(methylamino)-4-oxo-butanoic acid
(2R)-2-Amino-4-(methylamino)-4-oxo-butanoic acid
Formel: C₅H₁₀N₂O₃
Lager Temperatur: Raumtemperatur
MDL Number: MFCD04114468
CAS-Nummer: 7175-34-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR