(R)-4-Benzyl-2-(hydroxymethyl)morpholine

Lieferant: Apollo Scientific

Synonyms: (R)-(4-Benzylmorpholin-2-yl)methanol

OR471038-5G
APOSOR471038-5GEA 213 CHF
APOSOR471038-5G
(R)-4-Benzyl-2-(hydroxymethyl)morpholine
(R)-4-Benzyl-2-(hydroxymethyl)morpholine
Formel: C₁₂H₁₇NO₂
MW: 207.269 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD11848685
CAS-Nummer: 943442-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR