2-Bromo-7-methoxynaphthalene

Lieferant: Apollo Scientific

OR470815-1G
APOSOR470815-1GEA 145 CHF
APOSOR470815-1G
2-Bromo-7-methoxynaphthalene
2-Bromo-7-methoxynaphthalene
Formel: C₁₁H₉BrO
MW: 237,093 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD17012538
CAS-Nummer: 200875-36-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR