Methyl 1-(2-Methyl-3-nitrophenyl)-4-oxocyclohexanecarboxylate

Lieferant: Apollo Scientific

OR470537-5G
APOSOR470537-5GEA 1350 CHF
APOSOR470537-5G
Methyl 1-(2-Methyl-3-nitrophenyl)-4-oxocyclohexanecarboxylate
Methyl 1-(2-Methyl-3-nitrophenyl)-4-oxocyclohexanecarboxylate
Formel: C₁₅H₁₇NO₅
Lager Temperatur: Raumtemperatur
MDL Number: MFCD22423144
CAS-Nummer: 1385694-51-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR