Ethyl 3-Bromo-5-chloroindole-1-carboxylate

Lieferant: Apollo Scientific

OR470494-5G
APOSOR470494-5GEA 855 CHF
APOSOR470494-5G
Ethyl 3-Bromo-5-chloroindole-1-carboxylate
Ethyl 3-Bromo-5-chloroindole-1-carboxylate
Formel: C₁₁H₉BrClNO₂
MW: 302,552 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD22205871
CAS-Nummer: 1375064-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR