(5S,18S)-1,22-Diamino-7,16-dioxo-6,8,15,17-tetraazadocosane-5,18-dicarboxylic acid

Lieferant: Apollo Scientific

OR470114-1G
APOSOR470114-1GEA 477 CHF
APOSOR470114-1G
(5S,18S)-1,22-Diamino-7,16-dioxo-6,8,15,17-tetraazadocosane-5,18-dicarboxylic acid
(5S,18S)-1,22-Diamino-7,16-dioxo-6,8,15,17-tetraazadocosane-5,18-dicarboxylic acid
Formel: C₂₀H₄₀N₆O₆
Lager Temperatur: Raumtemperatur
MDL Number: MFCD16883073
CAS-Nummer: 1417789-80-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR