Methyl 2-Bromo-5-hydroxy-4-methylbenzoate

Lieferant: Apollo Scientific

OR46691-100MG OR46691-250MG OR46691-500MG
APOSOR46691-100MGEA 283 CHF
APOSOR46691-100MG APOSOR46691-250MG APOSOR46691-500MG
Methyl 2-Bromo-5-hydroxy-4-methylbenzoate
Methyl 2-Bromo-5-hydroxy-4-methylbenzoate
Formel: C₉H₉BrO₃
MW: 245.07 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD28976442
CAS-Nummer: 87808-27-3
New Product

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR