2-[(Dimethylamino)methylene]indan-1-one

Lieferant: Apollo Scientific
Achtung

OR46645-1G
APOSOR46645-1GEA 529 CHF
APOSOR46645-1G
2-[(Dimethylamino)methylene]indan-1-one
2-[(Dimethylamino)methylene]indan-1-one
Formel: C₁₂H₁₃NO
MW: 187,238 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00219908
CAS-Nummer: 150542-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR