3-Iodophenacyl bromide

Lieferant: Apollo Scientific
Gefahr

Synonyms: 2-Bromo-1-(3-iodophenyl)ethanone, 2-Bromo-1-(3-iodophenyl)ethan-1-one

OR45138-1G OR45138-250MG
APOSOR45138-1GEA 610 CHF
APOSOR45138-1G APOSOR45138-250MG
3-Iodophenacyl bromide
3-Iodophenacyl bromide
Formel: C₈H₆BrIO
MW: 324.941 g/mol
Melting Pt: 62…65 °C
Lager Temperatur: Kühlschrank
MDL Number: MFCD06666937
CAS-Nummer: 61858-38-6
UN: 3261
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR