2-Methylpyrrolidine-1,2-dicarboxylic acid-1-tert-butyl ester

Lieferant: Apollo Scientific
Achtung

Synonyms: 1-Boc-2-methyl-2-pyrrolidinecarboxylic acid, 1-Boc-2-methyl-DL-proline

OR451178-1G OR451178-250MG OR451178-5G
APOSOR451178-1GEA 119 CHF
APOSOR451178-1G APOSOR451178-250MG APOSOR451178-5G
2-Methylpyrrolidine-1,2-dicarboxylic acid-1-tert-butyl ester
2-Methylpyrrolidine-1,2-dicarboxylic acid-1-tert-butyl ester
Formel: C₁₁H₁₉NO₄
MW: 229.28 g/mol
Melting Pt: 104…106 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD03095494
CAS-Nummer: 203869-80-1

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR