5-Chinolincarbonsäure

Lieferant: Apollo Scientific
Achtung

OR451147-5G OR451147-1G OR451147-10G
APOSOR451147-5GEA 80.1 CHF
APOSOR451147-5G APOSOR451147-1G APOSOR451147-10G
5-Chinolincarbonsäure
5-Chinolincarbonsäure
Formel: C₁₀H₇NO₂
MW: 173.17 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00047608
CAS-Nummer: 7250-53-5

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR