L-Cyclobutylalanine

Lieferant: Apollo Scientific

OR451129-1G
APOSOR451129-1GEA 305 CHF
APOSOR451129-1G
L-Cyclobutylalanine
L-Cyclobutylalanine
Formel: C₇H₁₃NO₂
MW: 143.184 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD09836126
CAS-Nummer: 1201593-65-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR