N-tert-Butylhydroxylamine

Lieferant: Apollo Scientific

Synonyms: N-(tert-Butyl)hydroxylamine

OR45111-250MG
APOSOR45111-250MGEA 219 CHF
APOSOR45111-250MG
N-tert-Butylhydroxylamine
N-tert-Butylhydroxylamine
Formel: C₄H₁₁NO
Melting Pt: 59…65 °C
Dichte: 0.88 g/cm³ (25 °C)
Lager Temperatur: Kühlschrank
MDL Number: MFCD06653532
CAS-Nummer: 16649-50-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR