Boc-3-Cyclobutylalanine

Lieferant: Apollo Scientific

Synonyms: 2-((tert-Butoxycarbonyl)amino)-3-cyclobutylpropanoic acid

OR451070-5G
APOSOR451070-5GEA 985 CHF
APOSOR451070-5G
Boc-3-Cyclobutylalanine
Boc-3-Cyclobutylalanine
Formel: C₁₂H₂₁NO₄
MW: 243.299 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD09264344
CAS-Nummer: 565456-75-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR