1-(Ethylsulfonyl)piperazin 95%

Lieferant: Apollo Scientific
Achtung

OR4140-1G
APOSOR4140-1GEA 26.2 CHF
APOSOR4140-1G
1-(Ethylsulfonyl)piperazin 95%
1-(Ethylsulfonyl)piperazin
Formel: C₆H₁₄N₂O₂S
MW: 178,25 g/mol
Melting Pt: 69…72 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD02093528
CAS-Nummer: 62937-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR