Dicyclohexyl(2',4',6'-triisopropyl-2-biphenylyl)phosphin 97%

Lieferant: Apollo Scientific
Achtung

Synonyms: XPhos, 2-(Dicyclohexylphosphino)-2',4',6'-tri-i-propyl-1,1'-biphenyl, 2-Dicyclohexylphosphino-2',4',6'-triisopropylbiphenyl

OR40651-5G OR40651-25G OR40651-100G
APOSOR40651-5GEA 26.2 CHF
APOSOR40651-5G APOSOR40651-25G APOSOR40651-100G
Dicyclohexyl(2',4',6'-triisopropyl-2-biphenylyl)phosphin 97%
2-(Dicyclohexylphosphino)-2',4',6'-triisopropyl-1,1'-biphenyl
Formel: C₃₃H₄₉P
Melting Pt: 185…190 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD04117682
CAS-Nummer: 564483-18-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR