3-Chloro-2-nitrobenzyl bromide 95+%

Lieferant: Apollo Scientific
Gefahr

Synonyms: 1-(Bromomethyl)-3-chloro-2-nitrobenzene

OR401019-250MG
APOSOR401019-250MGEA 107 CHF
APOSOR401019-250MG
3-Chloro-2-nitrobenzyl bromide 95+%
3-Chloro-2-nitrobenzyl bromide
Formel: C₇H₅BrClNO₂
Lager Temperatur: Raumtemperatur
MDL Number: MFCD17168787
CAS-Nummer: 56433-00-2
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR