2-Chloro-6-iodobenzyl bromide ≥95%

Lieferant: Apollo Scientific

OR400751-250MG OR400751-1G
APOSOR400751-250MGEA 227 CHF
APOSOR400751-250MG APOSOR400751-1G
2-Chloro-6-iodobenzyl bromide ≥95%
2-Chloro-6-iodobenzyl bromide
Formel: C₇H₅BrClI
MW: 331.37606 g/mol
MDL Number: MFCD18393366
CAS-Nummer: 1035263-31-0
UN: 3261

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR