3-Bromo-2-iodobenzaldehyde 95%

Lieferant: Apollo Scientific
Achtung

OR400746-1G OR400746-250MG OR400746-5G
APOSOR400746-1GEA 688 CHF
APOSOR400746-1G APOSOR400746-250MG APOSOR400746-5G
3-Bromo-2-iodobenzaldehyde 95%
3-Bromo-2-iodobenzaldehyde
Formel: C₇H₄BrIO
Lager Temperatur: Raumtemperatur
MDL Number: MFCD18391618
CAS-Nummer: 1261860-40-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR