2-Bromo-5-nitrobenzenesulphonyl chloride

Lieferant: Apollo Scientific

OR400707-1G OR400707-250MG OR400707-5G
APOSOR400707-1GEA 563 CHF
APOSOR400707-1G APOSOR400707-250MG APOSOR400707-5G
2-Bromo-5-nitrobenzenesulphonyl chloride
2-Bromo-5-nitrobenzenesulphonyl chloride
Formel: C₆H₃BrClNO₄S
Lager Temperatur: Raumtemperatur
MDL Number: MFCD06254509
CAS-Nummer: 98130-55-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR