3-Bromo-4-nitrobenzenesulphonyl chloride

Lieferant: Apollo Scientific
Gefahr

OR400681-1G OR400681-250MG OR400681-5G
APOSOR400681-1GEA 557 CHF
APOSOR400681-1G APOSOR400681-250MG APOSOR400681-5G
3-Bromo-4-nitrobenzenesulphonyl chloride
3-Bromo-4-nitrobenzenesulphonyl chloride
Formel: C₆H₃BrClNO₄S
Lager Temperatur: Raumtemperatur
MDL Number: MFCD12197192
CAS-Nummer: 1181458-26-3
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR