2-Brom-6-chloranisol

Lieferant: Apollo Scientific
Achtung

Synonyms: 1-Bromo-3-chloro-2-methoxybenzene

OR400467-1G
APOSOR400467-1GEA 61 CHF
APOSOR400467-1G
2-Brom-6-chloranisol
2-Brom-6-chloranisol
Formel: C₇H₆BrClO
MW: 221.48 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD15143586
CAS-Nummer: 174913-10-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR