5-Bromo-2-methoxybenzyl bromide

Lieferant: Apollo Scientific

Synonyms: 4-Bromo-2-(bromomethyl)-1-methoxybenzene

OR400106-1G
APOSOR400106-1GEA 126 CHF
APOSOR400106-1G
5-Bromo-2-methoxybenzyl bromide
5-Bromo-2-methoxybenzyl bromide
Formel: C₈H₈Br₂O
MW: 279.957 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD07324845
CAS-Nummer: 184970-28-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR