5-Brom-2-chlor-4-(dimethylamino)pyrimidin

Lieferant: Apollo Scientific
Achtung

OR3707-1G
APOSOR3707-1GEA 55.7 CHF
APOSOR3707-1G
5-Brom-2-chlor-4-(dimethylamino)pyrimidin
5-Bromo-2-chloro-4-(dimethylamino)pyrimidine
Formel: C₆H₇BrClN₃
Melting Pt: 62…64 °C
Lager Temperatur: Kühlschrank
CAS-Nummer: 57054-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR