6-Azaindol 99%

Lieferant: Apollo Scientific
Achtung

OR3694-5G OR3694-1G
APOSOR3694-5GEA 78.4 CHF
APOSOR3694-5G APOSOR3694-1G
6-Azaindol 99%
6-Azaindol
Formel: C₇H₆N₂
Lager Temperatur: Kühlschrank
MDL Number: MFCD01686899
CAS-Nummer: 271-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR