2-Methoxypyrimidin-5-ylboronic acid, pinacol ester 95+%

Lieferant: Apollo Scientific

Synonyms: 2-Methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine, 2-Methoxypyrimidin-5-ylboronic acid pinacol ester

OR361403-250MG
APOSOR361403-250MGEA 73.1 CHF
APOSOR361403-250MG
2-Methoxypyrimidin-5-ylboronic acid, pinacol ester 95+%
2-Methoxypyrimidin-5-ylboronic acid, pinacol ester
Formel: C₁₁H₁₇BN₂O₃
MW: 236,075 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD13182190
CAS-Nummer: 1052686-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR