1,3,5-Trimethyl-1H-pyrazol-4-boronsäure 96%

Lieferant: Apollo Scientific

Synonyms: (1,3,5-Trimethyl-1H-pyrazol-4-yl)boronic acid

OR360564-1G
APOSOR360564-1GEA 64.5 CHF
APOSOR360564-1G
1,3,5-Trimethyl-1H-pyrazol-4-boronsäure 96%
1,3,5-Trimethyl-1H-pyrazole-4-boronic acid
Formel: C₆H₁₁BN₂O₂
MW: 153,9800 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD11183310
CAS-Nummer: 847818-62-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR