2-Methoxy-4-methyl-5-pyridinboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyms: 6-Methoxy-4-methylpyridin-3-ylboronic acid, 2-Methoxy-4-methylpyridine-5-boronic acid

OR360255-250MG
APOSOR360255-250MGEA 48.8 CHF
APOSOR360255-250MG
2-Methoxy-4-methyl-5-pyridinboronsäure
2-Methoxy-4-methyl-5-pyridinboronsäure
Formel: C₇H₁₀BNO₃
MW: 166,97 g/mol
Lager Temperatur: Kühlschrank
MDL Number: MFCD07368880
CAS-Nummer: 503184-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR