1-N-Boc-pyrrolidin-2-ylboronic acid 97%

Lieferant: Apollo Scientific

Synonyms: 1-N-Boc-Pyrrolidin-2-ylboronic acid, 2-Borono-1-pyrrolidinecarboxylic acid 1-(1,1-dimethylethyl) ester

OR360152-5G
APOSOR360152-5GEA 87 CHF
APOSOR360152-5G
1-N-Boc-pyrrolidin-2-ylboronic acid 97%
1-N-Boc-pyrrolidin-2-ylboronic acid
Formel: C₉H₁₈BNO₄
MW: 215,0545 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD02183523
CAS-Nummer: 149682-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR