Methyl 6-chloro-1H-indole-5-carboxylate 97%

Lieferant: Apollo Scientific

OR350504-250MG
APOSOR350504-250MGEA 48.8 CHF
APOSOR350504-250MG
Methyl 6-chloro-1H-indole-5-carboxylate 97%
Methyl 6-chloro-1H-indole-5-carboxylate
Formel: C₁₀H₈ClNO₂
MW: 209,629 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD11976069
CAS-Nummer: 162100-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR