1-N-Boc-4-(4-bromophenyl)piperidine

Lieferant: Apollo Scientific

Synonyms: 1-N-Boc-4-(4-Bromophenyl)piperidine

OR350304-5G
APOSOR350304-5GEA 126 CHF
APOSOR350304-5G
1-N-Boc-4-(4-bromophenyl)piperidine
1-N-Boc-4-(4-bromophenyl)piperidine
Formel: C₁₆H₂₂BrNO₂
MW: 340.2600 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD09878774
CAS-Nummer: 769944-78-7

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR