4-(Methylamino)-3-nitrobenzoesäure 98%

Lieferant: Apollo Scientific
Achtung

OR350255-100G OR350255-25G
APOSOR350255-100GEA 94 CHF
APOSOR350255-100G APOSOR350255-25G
4-(Methylamino)-3-nitrobenzoesäure 98%
4-(Methylamino)-3-nitrobenzoic acid
Formel: C₈H₈N₂O₄
MW: 196.1600 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00464322
CAS-Nummer: 41263-74-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR