3-Iod-1H-pyrazol

Lieferant: Apollo Scientific
Achtung

Synonyms: 3-Iodopyrazole

OR350207-25G OR350207-5G OR350207-10G
APOSOR350207-25GEA 180 CHF
APOSOR350207-25G APOSOR350207-5G APOSOR350207-10G
3-Iod-1H-pyrazol
3-Iodo-1H-pyrazole
Formel: C₃H₃IN₂
Lager Temperatur: Raumtemperatur
MDL Number: MFCD12022324
CAS-Nummer: 4522-35-4

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR