6-Phenyl-pyridazine-3-thiol

Lieferant: Apollo Scientific

OR346358-1G
APOSOR346358-1GEA 0 CHF
APOSOR346358-1G
6-Phenyl-pyridazine-3-thiol
6-Phenyl-pyridazine-3-thiol
Formel: C₁₀H₈N₂S
MW: 188,249 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD09039301
CAS-Nummer: 51841-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR