3-Bromo-1-methyl-1H-indole-2-carboxylic acid 95%

Lieferant: Apollo Scientific

OR33530-1G
APOSOR33530-1GEA 403 CHF
APOSOR33530-1G
3-Bromo-1-methyl-1H-indole-2-carboxylic acid 95%
3-Bromo-1-methyl-1H-indole-2-carboxylic acid
Formel: C₁₀H₈BrNO₂
MW: 254.09 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD11099656
CAS-Nummer: 499983-77-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR