2-Chloro-4-(methoxymethyl)-6-methylquinolin-7-amine, tech

Lieferant: Apollo Scientific

OR33173-1G
APOSOR33173-1GEA 1880 CHF
APOSOR33173-1G
2-Chloro-4-(methoxymethyl)-6-methylquinolin-7-amine, tech
2-Chloro-4-(methoxymethyl)-6-methylquinolin-7-amine
Formel: C₁₂H₁₃ClN₂O
MW: 236,697 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD03848702
CAS-Nummer: 866134-16-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR