3-(Dimethylamino)-1-(pyridin-3-yl)propan-1-ol, tech

Lieferant: Apollo Scientific

OR33015-1G
APOSOR33015-1GEA 1880 CHF
APOSOR33015-1G
3-(Dimethylamino)-1-(pyridin-3-yl)propan-1-ol, tech
3-(Dimethylamino)-1-(pyridin-3-yl)propan-1-ol
Formel: C₁₀H₁₆N₂O
MW: 180,247 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD02187195
CAS-Nummer: 478063-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR