2-(4-Methoxyphenyl)isonicotinonitril 95%

Lieferant: Apollo Scientific

Synonyms: 2-(4-Methoxyphenyl)pyridine-4-carbonitrile

OR32701-1G
APOSOR32701-1GEA 805 CHF
APOSOR32701-1G
2-(4-Methoxyphenyl)isonicotinonitril 95%
2-(4-Methoxyphenyl)isonicotinonitril
Formel: C₁₃H₁₀N₂O
Lager Temperatur: Raumtemperatur
MDL Number: MFCD07366546
CAS-Nummer: 885949-66-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR