trans-DL-1,2-Cyclopentanedicarboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

OR323252-1G
APOSOR323252-1GEA 76.6 CHF
APOSOR323252-1G
trans-DL-1,2-Cyclopentanedicarboxylic acid 97%
trans-DL-1,2-Cyclopentanedicarboxylic acid
Formel: C₇H₁₀O₄
MW: 158.153 g/mol
Melting Pt: 163…165 °C
Lager Temperatur: Raumtemperatur
CAS-Nummer: 1461-97-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR