2-Methylquinolin-3-ol 95%

Lieferant: Apollo Scientific

OR32268-1G
APOSOR32268-1GEA 1880 CHF
APOSOR32268-1G
2-Methylquinolin-3-ol 95%
2-Methylquinolin-3-ol
Formel: C₁₀H₉NO
MW: 159.185 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD02239638
CAS-Nummer: 613-19-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR