Methyl 4-(cyanoacetyl)benzoate

Lieferant: Apollo Scientific

OR322117-1G
APOSOR322117-1GEA 76.6 CHF
APOSOR322117-1G
Methyl 4-(cyanoacetyl)benzoate
Methyl 4-(cyanoacetyl)benzoate
Formel: C₁₁H₉NO₃
MW: 203,194 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00015378
CAS-Nummer: 69316-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR