1,1-Dimethylbiguanid Hydrochlorid

Lieferant: Apollo Scientific
Achtung

Synonyms: AMPK Signaling Activator VII, Metformin, Metformin hydrochloride, 1,1-Dimethylbiguanide Hydrochloride

OR322044-100G OR322044-10G OR322044-25G
APOSOR322044-100GEA 74.9 CHF
APOSOR322044-100G APOSOR322044-10G APOSOR322044-25G
1,1-Dimethylbiguanid Hydrochlorid
1,1-Dimethylbiguanid Hydrochlorid
Formel: C₄H₁₂ClN₅
MW: 165.625 g/mol
Melting Pt: 221 °C
Dichte: 1.36 g/cm³ (25 °C)
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00012582
CAS-Nummer: 1115-70-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR