3-(2,3-Dihydro-1H-isoindol-2-yl)-3-(4-methoxyphenyl)propan-1-ol, tech

Lieferant: Apollo Scientific

OR32140-1G
APOSOR32140-1GEA 1880 CHF
APOSOR32140-1G
3-(2,3-Dihydro-1H-isoindol-2-yl)-3-(4-methoxyphenyl)propan-1-ol, tech
3-(2,3-Dihydro-1H-isoindol-2-yl)-3-(4-methoxyphenyl)propan-1-ol
Formel: C₁₈H₂₁NO₂
MW: 283,365 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD04124253
CAS-Nummer: 860644-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR