2-[(4-Chlorophenyl)methyl]-1-azabicyclo[2.2.2]octan-3-one, tech

Lieferant: Apollo Scientific

OR32110-1G
APOSOR32110-1GEA 1880 CHF
APOSOR32110-1G
2-[(4-Chlorophenyl)methyl]-1-azabicyclo[2.2.2]octan-3-one, tech
2-[(4-Chlorophenyl)methyl]-1-azabicyclo[2.2.2]octan-3-one
Formel: C₁₄H₁₆ClNO
MW: 249.736 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD02082068
CAS-Nummer: 112628-51-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR