1,2-Dibrom-4-nitrobenzol

Lieferant: Apollo Scientific
Achtung

OR321004-25G
APOSOR321004-25GEA 115 CHF
APOSOR321004-25G
1,2-Dibrom-4-nitrobenzol
1,2-Dibromo-4-nitrobenzene
Formel: C₆H₃Br₂NO₂
MW: 280,9 g/mol
Melting Pt: 53…55 °C
Lager Temperatur: Raumtemperatur
MDL Number: MFCD00055373
CAS-Nummer: 5411-50-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR