(2-Ethylhexyl)magnesium bromide 1 M in Diethylether

Lieferant: Apollo Scientific

Synonyms: 2-Ethylhexylmagensium bromide

OR320020-100ML
APOSOR320020-100MLEA 227 CHF
APOSOR320020-100ML
(2-Ethylhexyl)magnesium bromide 1 M in Diethylether
(2-Ethylhexyl)magnesium bromide
Formel: C₈H₁₇BrMg
MW: 217.43 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD02260220
CAS-Nummer: 90224-21-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR