(1-Methylcyclobutyl)methanamine hydrochloride

Lieferant: Apollo Scientific
Achtung

OR317109-100MG OR317109-250MG
APOSOR317109-100MGEA 29.7 CHF
APOSOR317109-100MG APOSOR317109-250MG
(1-Methylcyclobutyl)methanamine hydrochloride
(1-Methylcyclobutyl)methanamine hydrochloride
Formel: C₆H₁₄ClN
MW: 135.64 g/mol
Lager Temperatur: Raumtemperatur
MDL Number: MFCD18074449
CAS-Nummer: 1245647-53-3

Order Now

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR