N-Boc-L-prolinal 97%

Lieferant: Apollo Scientific
Achtung

Synonyms: (S)-1-Boc-2-Formylpyrrolidine

OR316013-25G
APOSOR316013-25GEA 71.5 CHF
APOSOR316013-25G
N-Boc-L-prolinal 97%
N-Boc-L-prolinal
Formel: C₁₀H₁₇NO₃
MW: 199.25 g/mol
Siedepunkt: 211 °C
Dichte: 1.063 g/cm³ (25 °C)
Flash-Pt: 57.2 °C
Lager Temperatur: Gefrierschrank
MDL Number: MFCD00274186
CAS-Nummer: 69610-41-9
UN: 1989
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR